Order Entry
Denmark
ContactUsLinkComponent
HSQ Kit. HPLC performance qualification test kit complete with HPLC column NIST traceable solutions accessories and code to download the most current software and instructions from our website. . 1 * 1 items
  MICS49101-07
 :  MICROSOLV
undefined
HSQ Kit. HPLC performance qualification test kit complete with HPLC column NIST traceable solutions accessories and code to download the most current software and instructions from our website. . 1 * 1 items
  MICS49101-07
 :  MICROSOLV
Frequently Bought Together