Order Entry
Denmark
ContactUsLinkComponent
Metabolomics Column Kit, Cogent Diamond Hydride 2.o and Cogent Bidentate C18 2.o. Both columns are 1.0mm x 100mm, 2.2um, 120A. 1 * 2 items
Catalog # MICS43200-10P-1
Supplier:  MICROSOLV
undefined
Metabolomics Column Kit, Cogent Diamond Hydride 2.o and Cogent Bidentate C18 2.o. Both columns are 1.0mm x 100mm, 2.2um, 120A. 1 * 2 items
Catalog # MICS43200-10P-1
Supplier:  MICROSOLV
Frequently Bought Together