Order Entry
Denmark
ContactUsLinkComponent
Cogent 2.o HPLC 5 Column Suite including Silica-C, Bidentate C8, Bidentate C18, UDC Cholesterol and Diamond Hydride 2.1mm x 100mm, 2 .2um 120A. 1 * 5 items
Catalog # MICS42100-10P-2
Supplier:  MICROSOLV
undefined
Cogent 2.o HPLC 5 Column Suite including Silica-C, Bidentate C8, Bidentate C18, UDC Cholesterol and Diamond Hydride 2.1mm x 100mm, 2 .2um 120A. 1 * 5 items
Catalog # MICS42100-10P-2
Supplier:  MICROSOLV
Frequently Bought Together