Multi-parameter meters, handheld, Eutech PC 450 / PD 450
Product Details & Documents
These portable meters either measure pH, mV, conductivity, TDS, salinity and temperature (PC 450) or pH, mV, DO and temperature (PD 450). They feature a durable waterproof and dustproof design with IP67 rating.
- Large, backlit display with intuitive user-friendly icons
- Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
- Automatic pH buffer recognition; up to 5- or 6-point calibration; calibration due alarm
- Stores up to 500 data sets; easy USB or RS232 output with data cables (cables not always included)
- Meets GLP standards: Built-in real-time clock stamps stored data and calibration data with time and date
- Can be used with two AA batteries or with optional universal power adapter (not included)
- Built-in stand with wall-mount option allows for easy bench top measurement or storage
PC 450 accepts a single pH/conductivity probe or separate pH and conductivity probes.
Leveringsinformation: PC 450 meter is supplied with combi pH/Con/ATC electrode and two AA batteries.
PC 450 Kit is supplied with double-junction pH electrode, conductivity cell (K = 1,0), Grip-Clip™, two AA batteries, hard carry case, USB cable, and calibration solutions.
PC 450 Combi Kit is supplied with combi pH/Con/ATC electrode, two AA batteries, hard carry case, USB cable, and calibration solutions.
PD 450 Kit is supplied with pH electrode, RDO electrode with sensor cap, two AA batteries, hard carry case, USB cable, RDO stainless steel weight/guard, RDO calibration chamber and calibration solutions.
Documents
eutech-150-450-series-family-brochure
Product Literature
Specifications
- Description:Multi-parameter meter (pH / mV / conductivity / TDS / salinity / temperature), PC 450, kit with pH electrode and conductivity cell
- Seq:02
- Compliance:IP67
- DO concentration resolution:-
- Conductivity resolution:0,01 µS - 0,1 mS
- DO accuracy:-
- Data logging capacity:Up to 500 data sets
- Power supply:Two AA batteries (included), AC adapter (optional)
- mV calibration:±200
- mV resolution:0,1 ±999,9; 1 beyond
- TDS accuracy:±1 % full scale
- Conductivity accuracy:±1 % full scale
- DO calibration:-
- Conductivity calibration:Auto/manual; one point per range
- W×D×H:200×83×57 mm
- Temperature range:−10,0...+110,0 °C
- Temperature calibration:Offset in 0,1 increments, ±10 °C
- mV range:±2000
- TDS calibration:Manual; 1 point/range
- pH range:−2,00...+16,00
- Temperature compensation:Automatic or manual from −10 to 110 °C
- pH calibration:Up to 6 points: USA, NIST, DIN or custom
- Temperature resolution:0,1 °C
- DO concentration:-
- TDS resolution:0,01 ppm - 0,1 ppt
- Temperature accuracy:±0,5 °C
- Weight:0,5 kg
- Cat. No.:665-0488
- pH accuracy:±0,01
- pH resolution:0,01
- Model:PC 450
- Conductivity range:0 - 200 mS
- Outputs:USB, RS232
- TDS range:0 - 200 ppt (TDS factor 1,0)
- mV accuracy:±0,2+2 LSD or +0,5 % of reading; whichever is greater
- DO saturation:-
- Display:Custom LCD with backlight
Accessories
Product Information
- Catalog #pHTemp.AvailabilityPrice
- pH electrode, double junction, sealedpH electrode, double junction, sealedEUTEECFC7252201B1 - 130...+80 °CLog in for availability1 * 1 Items - 993,00 kr
Product Information
- Catalog #ConductivityCell constantAvailabilityPrice
- Conductivity/ATC cellConductivity/ATC cellEUTECONSEN91W0 - 150 mS1,0 cm⁻¹Log in for availability1 * 1 Items - 1.990,00 kr
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



