HT100 instrument with full touchscreen colour graphic display. Includes PK19M Penetration probe, Protective Boot and Micro USB cable. Free software via weblink download, see Quick Reference Guide supplied when ordered. -73°C to +537°C / -99°F to +999 1 * 1 items
Catalog # COMAHT100/PK19
Supplier: COMARK
Frequently Bought Together