Order Entry
Denmark
ContactUsLinkComponent
Bluetooth OEM App Applications, Waterproof Pocketherm digital thermometer (T-thermocouple)-58°F to +482°F 1 * 1 items
Catalog # COMABT250
Supplier:  COMARK
Bluetooth OEM App Applications, Waterproof Pocketherm digital thermometer (T-thermocouple)-58°F to +482°F 1 * 1 items
Catalog # COMABT250
Supplier:  COMARK
Frequently Bought Together