Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cleaning coupons
Cleaning coupons
Cleaning coupon, PTFE, 100×100 mm
Catalog # SAMP8644T-100
Supplier:   Sampling Systems
Cleaning coupons
Catalog # SAMP8644T-100
Supplier:  Sampling Systems
Cleaning coupons
Cleaning coupons
Cleaning coupon, PTFE, 100×100 mm
Price
1.310,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These cleaning validation coupons are available in all popular materials and various sizes.
Specifications
  • Description:
    Cleaning coupon, PTFE
  • L×W:
    100×100 mm
  • Seq:
    06
Anbefalinger