Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant PDGF-BB

100 µG
Catalog # STMC78097.1
Marketsource
Human Recombinant PDGF-BB
Catalog # STMC78097.1
Marketsource
CAS Number:  
Human Recombinant PDGF-BB
Human Recombinant PDGF-BB

100 µG

CAS Number:  
Price
6.338,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Platelet-derived growth factor (PDGF) is a dimeric glycoprotein consisting of two disulfide bridge stabilized polypeptide chains, A and B, which are assembled as heterodimers (PDGF-AB) or homodimers (PDGF-AA and PDGF-BB) (Fretto et al.; Westermark and Heldin). PDGF signals through the receptor tyrosine kinases PDGFRalpha and PDGFRbeta. It has been shown that PDGF-induced migration involves signaling pathways involving MEK/ERK, EGFR, Src and PI3K/AKT (Kim et al.). PDGF is a potent mitogen for cells of mesenchymal origin- like fibroblasts, glial cells, and vascular smooth muscle cells. PDGF has been implicated in pathogenesis of atherosclerosis, glomerulonephritis, cancer, and in the contraction of vascular smooth muscle cells of rat aortic tissues (Fretto et al.; Sachinidis et al.). PDGF-BB is secreted by osteoblasts to induce mesenchymal stem cell migration and angiogenesis. It has also been shown that PDGF-BB is secreted by preosteoclasts during bone modeling and remodeling to induce angiogenesis and thus proper osteogenesis (Xie et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    STMC78097.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    PDGF-BB
  • Purity:
    ≥95%
  • Species:
    Human

Specifications