Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant BAFF

50 µG
Catalog # STMC78002.1
Marketsource
Human Recombinant BAFF
Catalog # STMC78002.1
Marketsource
CAS Number:  
Human Recombinant BAFF
Human Recombinant BAFF

50 µG

CAS Number:  
Price
4.991,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

B cell activating factor (BAFF) is part of the tumor necrosis factor (TNF) superfamily and is expressed by monocytes, macrophages, dendritic cells, and neutrophils (Mackay and amp; Browning; Scapini et al.). It binds to 3 receptors: BAFF-R, TACI, and BCMA, all of which are expressed by B cells at various times in their development (Liu and amp; Davidson). BAFF-R signaling activates the NF-kB signaling pathway, and promotes B cell survival, Ig class switching, and antigen presentation (Bossen and amp; Schneider; Liu and amp; Davidson; Naradikian et al.). BAFF also has functions in other cell types: it promotes monocyte survival and activation, proinflammatory cytokine secretion, and differentiation into macrophages (Chang et al.), and co-stimulates anti-CD3-mediated activation of human T cells (Huard et al.; Mackay and amp; Leung).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    STMC78002.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    BAFF
  • Purity:
    <gt/>95%
  • Species:
    Human

Specifications