Order Entry
Denmark
Orders LinkContactUsLinkComponent
STEMdiff™ APEL™2 Medium
STEMdiff™ APEL™2 Medium
STEMdiff™ APEL™2 Medium, 500 mL
Catalog # STMC05275
Marketsource
STEMdiff™ APEL™2 Medium
Catalog # STMC05275
Marketsource
CAS Number:  
STEMdiff™ APEL™2 Medium
STEMdiff™ APEL™2 Medium
STEMdiff™ APEL™2 Medium, 500 mL
CAS Number:  
Price
7.842,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents
Defined, animal component-free medium for differentiation of human ES and iPS cells to multiple lineages.
STEMdiff™ APEL™ 2 Medium is a fully defined, serum-free and animal component-free medium for the differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. It is based on the APEL formulation published by Dr. Andrew Elefanty and lacks undefined components such as protein-free hybridoma medium. STEMdiff™ APEL™ 2 can be used in adherent or embryoid body (EB)-based protocols, such as with AggreWell™. It can be used with a variety of different induction factors or cytokines to support differentiation along ectoderm, mesoderm and endoderm lineages.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    STEMdiff™ APEL™2 Medium