Sodium azide 99+%
Plastic bottle, 500 g
Catalog # J21610.36
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Sodium triazide
- CAS Number:26628-22-8
- Formula:NaN₃
- MDL Number:MFCD00003536
- Molecular Weight:65.01 g/mol
- Merck Index:14,08581
- Melting Pt:∼275 °C (dec.)
- Density:1.846 g/cm³
- Storage Temperature:Ambient
- EINECS:247-852-1
- UN:1687
- ADR:6.1,II
Documents
Specifications
Specification Test Results
- Form:White crystalline powder
- Assay (Unspecified):≥99.0%
- Loss on Drying:≤0.5%
Specifications
- Pack type:Plastic bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



