Order Entry
Denmark
ContactUsLinkComponent
WHEN CLEANING CONES WHICH HAVE A SCREW THREAD, IT IS IMPORTANT THAT THE THREAD IS NOT CONTACTED BY ANY CORROSIVE SOLUTION. IF THE TH READ GETS CORRODED, THE CONE MAY NOT SEAL CORRECTLY OR IT MAY BOND TO THE BASE AND BE DIFFICULT TO REMOVE. AND WITH P 1 * 1 items
Catalog # PERCN8145319
Supplier:  PerkinElmer
WHEN CLEANING CONES WHICH HAVE A SCREW THREAD, IT IS IMPORTANT THAT THE THREAD IS NOT CONTACTED BY ANY CORROSIVE SOLUTION. IF THE TH READ GETS CORRODED, THE CONE MAY NOT SEAL CORRECTLY OR IT MAY BOND TO THE BASE AND BE DIFFICULT TO REMOVE. AND WITH P 1 * 1 items
Catalog # PERCN8145319
Supplier:  PerkinElmer
Frequently Bought Together