Polysorbate 20, J.T.Baker®
Glass bottle, amber, narrow mouth, PVC-coated, 4 L
Supplier: Avantor
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Density:1.1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:500-018-3
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Identification (by IR):Passes Test
Specifications
- Pack type:Glass bottle, amber, narrow mouth, PVC-coated
- Pk:4 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



