Order Entry
Denmark
Orders LinkContactUsLinkComponent
4-Methylmorpholine for synthesis

4-Methylmorpholine for synthesis

Glass bottle, 250 ml
Catalog # MERC8.05894.0250
Flammable
Corrosive
Irritant
Supplier:   Merck
4-Methylmorpholine for synthesis
Catalog # MERC8.05894.0250
Supplier:  Merck
CAS Number:  109-02-4
4-Methylmorpholine for synthesis

Glass bottle, 250 ml

CAS Number:  109-02-4
Price
246,00 kr
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  NMM, N-Methylmorpholine
  • CAS Number:109-02-4
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00006175
  • Molecular Weight:101.15 g/mol
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–66 °C
  • Density:0.922 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:203-640-0
  • ADR:3 (8),II
  •  :M-Clarity™ quality level = MQ200
  • UN:2535

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml

Specifications