Order Entry
Denmark
Orders LinkContactUsLinkComponent
Bottles, narrow neck, with thread, SYSTEM PLUS® 32
Bottles, narrow neck, with thread, SYSTEM PLUS® 32
1100 ml, DIN 32
Catalog # TOUR5243S1
Supplier:   TOURNAIRE-EMBALLAGE
Bottles, narrow neck, with thread, SYSTEM PLUS® 32
Catalog # TOUR5243S1
Bottles, narrow neck, with thread, SYSTEM PLUS® 32
Bottles, narrow neck, with thread, SYSTEM PLUS® 32
1100 ml, DIN 32
Price
113,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Pure aluminium.
  • Unbreakable, odorless
  • Very good temperature resistance
  • Leakproof with tamper-evident screw caps (must be odered separately)
  • UN-approved
Suitable for storage and transport of highly aggessive liquids.
Specifications
  • Thread:
    DIN 32
  • Capacity:
    1100 ml
  • Seq:
    04