Order Entry
Denmark
ContactUsLinkComponent
4 port 2-pos valve, u-electric, 1/16*.25mm, 50C/10000psi liq, N60C/E3, Universal electric actuator, interface: TTL (contact closure) 1 * 1 items
Catalog # VICIC82H-1674EUH
Supplier:  VICI
4 port 2-pos valve, u-electric, 1/16*.25mm, 50C/10000psi liq, N60C/E3, Universal electric actuator, interface: TTL (contact closure) 1 * 1 items
Catalog # VICIC82H-1674EUH
Supplier:  VICI
Frequently Bought Together