Order Entry
Denmark
ContactUsLinkComponent
The MiniDigi 1B is a two channel Digitizer for use with included Axoscope software acting as a simple digitial chart recorder. This can be used to monitor membrane parameters in real time while recording. Included is the MiniDigi 1B instrument and 1 * 1 items
Catalog # MLDVMINIDIGI1B
Supplier:  Molecular Devices
The MiniDigi 1B is a two channel Digitizer for use with included Axoscope software acting as a simple digitial chart recorder. This can be used to monitor membrane parameters in real time while recording. Included is the MiniDigi 1B instrument and 1 * 1 items
Catalog # MLDVMINIDIGI1B
Supplier:  Molecular Devices
Frequently Bought Together