Order Entry
Denmark
ContactUsLinkComponent
H2O2 Indicator Strip, Transitions Red to Blue Type 4 101 mm (4 ) x 16 mm (0.625 ) 1 * 250 items
Catalog # NAMSGPS-250R
Supplier:  CROSSTEX
H2O2 Indicator Strip, Transitions Red to Blue Type 4 101 mm (4 ) x 16 mm (0.625 ) 1 * 250 items
Catalog # NAMSGPS-250R
Supplier:  CROSSTEX