Order Entry
Denmark
Orders LinkContactUsLinkComponent
Calorimeters, C 200
C 200 H automated, water handling
Catalog # IKAC10002387
Supplier:   IKA
Calorimeters, C 200
Catalog # IKAC10002387
Supplier:  IKA
Calorimeters, C 200
Calorimeters, C 200
C 200 H automated, water handling
Price
kr269,530.00
Log in for availability
1 * 1 Items
Product Details & Documents
Compact and space-saving semi automated calorimeter to determine calorific values of liquid and solid samples.
  • Manual oxygen filling and venting, manual water preparation
  • The C 5010 decomposition vessel can be equipped to use C 14 disposable crucible
  • Halogen resistant vessel for quantitative decomposition of halogens and sulphur
  • Includes external power pack, complies with all global voltages from 100 to 240 V AC, 50/60 Hz; calorimeter is powered with a low operating voltage of 24 V DC
  • Suitable for use with Calwin C 5040 calorimeter software: monitoring and visualisation of measuring procedure
Suitable for educational purposes (example technical schools, universities) and for commercial, industrial laboratories with a low number of samples. Four different test procedures isoperibol, dynamic, time-controlled (automatic ignition and calculation of calorific value) or manual user-dependent ignition and manual calculations allow ideal adaptation to various tasks.
 : Supplied with Basic device (measuring cell) C 200 incl. power pack and ignition adaptor, decomposition vessel C 5010 or decomposition vessel halogen resistant C 5012, conversion kit C 200.2, oxygen station C 248 and further consumables for installation and calibration.
Specifications
  • Description:
    C 200 H automated, water handling
  • Seq:
    03
  • Cat. No.:
    IKAC10002387
  • Duration of measurement:
    Isoperibolic: ~ 17 min
    Dynamic: ~ 8 min
    Manual: ~ 17 min
    Time controlled: ~ 14 min
  • Reproducibility (upon analysis with 1 g benzoic acid):
    0,1% RSD
  • Absorbed power:
    120 W
  • Permitted ambient temperature:
    20...25 °C
  • Oxygen working pressure:
    30 bar
  • Max. measurement range:
    40000 J
  • Permitted ambient relative humidity:
    80 %
  • Measuring modes:
    Isoperibolic, dynamic, manual, time controlled
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Quartz crucible, big, C 6, Ø 26×18 mm (C 5010.5 crucible pan is required for usage)
    Quartz crucible, big, C 6, Ø 26×18 mm (C 5010.5 crucible pan is required for usage)
    IKAC0355100
    Log in for availability
    1 * 1 Items - 455,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Crucible holder, big model, C 5010.5 (required with "C 6 Quartz crucible, big")
    Crucible holder, big model, C 5010.5 (required with "C 6 Quartz crucible, big")
    IKAC3055900
    Log in for availability
    1 * 1 Items - 2.110,00 kr
Product Information
  • Catalog #
    Type
    Temperature Range
    Temp. stability
    Availability
    Price
  • IKAA4173000
    RC 2 control
    −20...+40 °C
    ±0,05
    Log in for availability
    1 * 1 Items - 43.320,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Paraffin strips, especially for hard flammable, water containing samples, 45×3 mm
    Paraffin strips, especially for hard flammable, water containing samples, 45×3 mm
    IKAC3131100
    Log in for availability
    1 * 600 Items - 564,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Acetobutyrate capsules with known calorific value to fill in samples with volatile compounds, not hygroscopic, C 10
    Acetobutyrate capsules with known calorific value to fill in samples with volatile compounds, not hygroscopic, C 10
    IKAC0750000
    Log in for availability
    1 * 100 Items - 1.150,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Quartz dish, Ø 20 mm × 20,5 mm high, C 4
    Quartz dish, Ø 20 mm × 20,5 mm high, C 4
    IKAC1695500
    Log in for availability
    1 * 1 Items - 332,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Conversion kit, C 200.2, for using the halogen resistent decomposition vessel C 5012 with the C 200 standard package
    Conversion kit, C 200.2, for using the halogen resistent decomposition vessel C 5012 with the C 200 standard package
    IKAC4028800
    Log in for availability
    1 * 1 Items - 1.760,00 kr
Product Information
  • Catalog #
    Type
    Temperature Range
    Temp. stability
    Availability
    Price
  • IKAA4171000
    RC 2 basic
    −20...+40 °C
    ±0,05
    Log in for availability
    1 * 1 Items - 34.030,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Set of VA combustion crucibles, C 5 (a single crucible is allowed to be used for up to 25 combustions Ø 20 mm x 19,5 mm high)
    Set of VA combustion crucibles, C 5 (a single crucible is allowed to be used for up to 25 combustions Ø 20 mm x 19,5 mm high)
    IKAC1749500
    Log in for availability
    1 * 1 Each - 1.570,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Ignition wire, platinum, C 1.123
    Ignition wire, platinum, C 1.123
    IKAC4500200
    Log in for availability
    1 * 2 Items - 1.540,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Combustion bags, PE, with known calorific value, to fill in samples with a low calorific value, powdery and with low specific gravity, 70×40 mm, C 12 A
    Combustion bags, PE, with known calorific value, to fill in samples with a low calorific value, powdery and with low specific gravity, 70×40 mm, C 12 A
    IKAC2201500
    Log in for availability
    1 * 100 Items - 465,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • C 5010 Stainless steel decomposition vessel, standard with fix ignition wire, Can be converted to use the C 14 combustible crucible
    C 5010 Stainless steel decomposition vessel, standard with fix ignition wire, Can be converted to use the C 14 combustible crucible
    IKAC20128010
    Log in for availability
    1 * 1 Items - 40.860,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Benzoic acid tablet, blister package, 0,5 g per pellet, C 723 (with IKA-certified gross calorific value for Standard-calibrations according to DIN 51900)
    Benzoic acid tablet, blister package, 0,5 g per pellet, C 723 (with IKA-certified gross calorific value for Standard-calibrations according to DIN 51900)
    IKAC3243000
    Log in for availability
    1 * 1 Each - 1.530,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Attachment for combustible crucible C 14 (required when using the C 14 disposable crucible)
    Attachment for combustible crucible C 14 (required when using the C 14 disposable crucible)
    IKAC3016900
    Log in for availability
    1 * 1 Items - 2.860,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Ignition wire, spare, C 5010.3, for about 100 combustions each, used in C 5010 and C 7010 decomposition vessels (for C 200)
    Ignition wire, spare, C 5010.3, for about 100 combustions each, used in C 5010 and C 7010 decomposition vessels (for C 200)
    IKAC7122800
    Log in for availability
    1 * 5 Items - 612,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Ignition wire, platinum, C5012.3, for about 50 combustions each, used in C 5012, C 7012 and AOD 1.1 decomposition vessels
    Ignition wire, platinum, C5012.3, for about 50 combustions each, used in C 5012, C 7012 and AOD 1.1 decomposition vessels
    IKAC2994900
    Log in for availability
    1 * 2 Items - 1.700,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Benzoic acid NBS 39i, C 43, Standard Reference Material with certificate of the "National In-stitute of Standards & Technology ( NIST ), USA"
    Benzoic acid NBS 39i, C 43, Standard Reference Material with certificate of the "National In-stitute of Standards & Technology ( NIST ), USA"
    IKAC0750600
    Log in for availability
    1 * 30 g - 6.930,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Benzoic acid, BIG Package, C 723, weight 0,5 g per pellet, 450 pieces (9 packages with 50 pieces), blister packaging (with IKA-certified gross calorific value for Standard-calibrations according to DIN 51900)
    Benzoic acid, BIG Package, C 723, weight 0,5 g per pellet, 450 pieces (9 packages with 50 pieces), blister packaging (with IKA-certified gross calorific value for Standard-calibrations according to DIN 51900)
    IKAC3717400
    Log in for availability
    1 * 450 Items - 9.500,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Paraffin, liquid, 30 ml, C 17 (combustion aid for samples with high water content percentage, as well as bonding agent for strongly powder samples)
    Paraffin, liquid, 30 ml, C 17 (combustion aid for samples with high water content percentage, as well as bonding agent for strongly powder samples)
    IKAC3801200
    Log in for availability
    1 * 30 mL - 1.070,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Parafilm tape, C 16, for closing the crucible combustion aid for liquids with volatile components, 1000×50 mm
    Parafilm tape, C 16, for closing the crucible combustion aid for liquids with volatile components, 1000×50 mm
    IKAC3801100
    Log in for availability
    1 * 1 Items - 645,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Combustible crucible, C 14, acetobutyrate, specially created for samples that are hard to burn, required in combination with C 5010.4 crucible pan (for C 200)
    Combustible crucible, C 14, acetobutyrate, specially created for samples that are hard to burn, required in combination with C 5010.4 crucible pan (for C 200)
    IKAC7224500
    Log in for availability
    1 * 100 Items - 1.140,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Combustion bags, 40×35 mm, C 12
    Combustion bags, 40×35 mm, C 12
    IKAC2201400
    Log in for availability
    1 * 100 Items - 455,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Cotton thread, C 710.8, cut to length, thick, 500 pieces (for C 1 / C 6000)
    Cotton thread, C 710.8, cut to length, thick, 500 pieces (for C 1 / C 6000)
    IKAC4579900
    Log in for availability
    1 * 1 Items - 427,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Set of VA combustion crucibles, big, C 710.2
    Set of VA combustion crucibles, big, C 710.2
    IKAC1483500
    Log in for availability
    1 * 25 Items - 2.410,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Cotton thread, 500 pieces, cut to length, with known calorific value, C 710-4
    Cotton thread, 500 pieces, cut to length, with known calorific value, C 710-4
    IKAC1483700
    Log in for availability
    1 * 500 Items - 422,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Venting station, C 5030, to control the degassing process of the combustion gases after the decomposition, suitable for the C 5010 and C 5012 decomposition vessels (for C 200)
    Venting station, C 5030, to control the degassing process of the combustion gases after the decomposition, suitable for the C 5010 and C 5012 decomposition vessels (for C 200)
    IKAC7198000
    Log in for availability
    1 * 1 Items - 30.050,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Control standard, AOD 1.11, 50 ml, mineral oil with cetrificated Sulfur- and Chlorine-content, for checking the handling, AOD-decomposition and the respective peripheral detection devices
    Control standard, AOD 1.11, 50 ml, mineral oil with cetrificated Sulfur- and Chlorine-content, for checking the handling, AOD-decomposition and the respective peripheral detection devices
    IKAC3044000
    Log in for availability
    1 * 50 mL - 3.330,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Manual pellet press for powders; good combustible substances, C 21
    Manual pellet press for powders; good combustible substances, C 21
    IKAC1605300
    Log in for availability
    1 * 1 Items - 34.900,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Gelatine capsules, C 9
    Gelatine capsules, C 9
    IKAC0749900
    Log in for availability
    1 * 100 Items - 436,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • CalWin control and evaluation software, C 6040, for IKA® calorimeters
    CalWin control and evaluation software, C 6040, for IKA® calorimeters
    IKAC4040500
    Log in for availability
    1 * 1 Items - 26.380,00 kr