Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR® AVS TITRINORM Buffer Solutions, 20 °C

VWR® AVS TITRINORM Buffer Solutions, 20 °C

10,00, ±0,02, Carbonate, Yellow, ISO 17034, Plastic bottle, 1 L
Catalog # 85680.295
Supplier:   VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Catalog # 85680.295
Supplier:  VWR Chemicals
CAS Number:  
VWR® AVS TITRINORM Buffer Solutions, 20 °C
VWR® AVS TITRINORM Buffer Solutions, 20 °C

10,00, ±0,02, Carbonate, Yellow, ISO 17034, Plastic bottle, 1 L

CAS Number:  
Price
354,00 kr
Log in for availability
1 * 1 L
Product Details & Documents

Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.

  • Reference temperature 20 °C
  • True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 11,00); ±0,05 for pH 12,00 buffer
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website

Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 11,00 and 0,05 pH unit for pH 12,0.

Certifications: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.

Ordering information: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.

Packaging: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Carbonate
  • Pk:
    1 L
  • Colour:
    Yellow
  • pH accuracy:
    ±0,02
  • pH value:
    10,00
  • Accreditation:
    ISO 17034

Specifications