Order Entry
Denmark
ContactUsLinkComponent
CF™570 maleimide is a superior red fluorescent dye with a maleimide reactive group, which can be used to label a target molecule wit h a thiol group. CF™570 has the maximum absorption at 568 nm that is optimally matching the 568 nm line of the Ar-Kr 1 * 1 µmol
Catalog # 96015.
Supplier:  Biotium
CF™570 maleimide is a superior red fluorescent dye with a maleimide reactive group, which can be used to label a target molecule wit h a thiol group. CF™570 has the maximum absorption at 568 nm that is optimally matching the 568 nm line of the Ar-Kr 1 * 1 µmol
Catalog # 96015.
Supplier:  Biotium
Frequently Bought Together