Order Entry
Denmark
Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ IXSelect
Affinity chromatography columns, HiTrap™ IXSelect
HiTrap™ IXSelect, PP, 1 ml
Catalog # GEHE17-3714-11
Supplier:   Cytiva
Affinity chromatography columns, HiTrap™ IXSelect
Catalog # GEHE17-3714-11
Supplier:  Cytiva
Affinity chromatography columns, HiTrap™ IXSelect
HiTrap™ IXSelect, PP, 1 ml
Price
8.280,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
HiTrap™ IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.
  • IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams
Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
Specifications
  • Capacity:
    1 ml
  • Description:
    HiTrap™ IXSelect
  • Column hardware:
    PP