Order Entry
Denmark
ContactUsLinkComponent
This gene encodes a member of the nucleotide-sugar transporter family. The encoded protein is a multi-pass membrane protein. It tran sports UDP-galactose from the cytosol into Golgi vesicles, where it serves as a glycosyl donor for the generation of 1 * 100 µG
  BWRLBS8791
This gene encodes a member of the nucleotide-sugar transporter family. The encoded protein is a multi-pass membrane protein. It tran sports UDP-galactose from the cytosol into Golgi vesicles, where it serves as a glycosyl donor for the generation of 1 * 100 µG
  BWRLBS8791
Frequently Bought Together