Galactose Mutarotase is a member of the aldose epimerase family and is involved in hexose metabolism. Through its catalytic activity , Galactose Mutarotase converts ∫-aldose to å-aldose on several sugars, including D-glucose, L-arabinose and D-xylose 1 * 100 µG
Catalog # BWRLBS61444
Supplier: Bioworld Technology
Frequently Bought Together