Short-term tube, DT n-butanol 10/a, standard measuring ranges of 10 to 250 ppm and 250 to 2000 ppm 1 * 10 items
Catalog # DRAE8103861
Supplier: Dräger
Short-term tube, DT n-butanol 10/a, standard measuring ranges of 10 to 250 ppm and 250 to 2000 ppm 1 * 10 items
Catalog # DRAE8103861
Supplier: Dräger
Frequently Bought Together
Accessories for titrators
Supplier: Mettler - Toledo
VWR® Counter-Pen™ Cell Counter
Supplier: VWR Collection
VWR®, Accessories for Pocket Colorimeters, MCP 100 and MCT 100
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
VWR®, Portable Turbidity Meter
Supplier: VWR Collection
Titration beakers
Supplier: Mettler - Toledo
Quick connect Luer adapters, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Accessories for cell counter, EVE™
Supplier: NANOENTEK
Electrode stand with swivel arm, Eutech
Supplier: EUTECH
Accessories for hydrogen generator precision hydrogen trace
Supplier: PEAK SCIENTIFIC
Belüftetes Qtray mit Deckel & 48 Wellteiler, Polystyrol, 242x240x20m
Supplier: Molecular Devices
Plastic cuvette for LICO
Supplier: Hach
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
Accessories for cuvette tests
Supplier: Hach