0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Skim milk butyrometer 0-1%: 0.01 % Fat: 0 - 1 division: 0.01 milk: 10.73 - 11 ml 1 * 1 items
Catalog # GEAG03.0231
Supplier:
Gerber Instruments AG
Expand image
Skim milk butyrometer 0-1%: 0.01 % Fat: 0 - 1 division: 0.01 milk: 10.73 - 11 ml 1 * 1 items
Catalog # GEAG03.0231
Supplier:
Gerber Instruments AG
Frequently Bought Together
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
Supplier: VWR Collection
Measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Supplier: Brand
Graduated pipettes, with piston
Supplier: HIRSCHMANN
Bulb pipettes (one mark), BLAUBRAND®, class AS
Supplier: Brand
VWR® Volumetric Flasks, Class A, Glass, Amber, NS Plastic Stopper
Supplier: VWR Collection
Measuring cylinder for jolting volumeter
Supplier: GERBER FUNKE
Measuring cylinders, tall form, class B, borosilicate glass, SILBERBRAND ETERNA
Supplier: Brand
VWR® Bulb Pipettes, Two Marks, Class AS
Supplier: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
Supplier: VWR Collection
VWR®, Bulb Pipettes, One Mark, Class AS
Supplier: VWR Collection
VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection