Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dextrin

Dextrin

100 g
Catalog # TCIAD4657-100G
Supplier:   TCI
Dextrin
Catalog # TCIAD4657-100G
Supplier:  TCI
CAS Number:  9004-53-9
Dextrin

100 g

CAS Number:  9004-53-9
Price
173,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Dextrin
  • CAS Number:9004-53-9
  • Formula:(C₆H₁₂O₆)n
  • MDL Number:MFCD00081554
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,02953
  • EINECS:232-675-4
Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Drying loss:
    max. 10.0 %
  • Ignition residue (Sulfate):
    max. 0.5 %
  • Reducing suger:
    max. 5.0 %

Specifications

  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.