Order Entry
Denmark
Orders LinkContactUsLinkComponent
Crystal violet

Crystal violet

100 g
Catalog # TCIAC0428-100G
Corrosive
Irritant
Health
Environment
Supplier:   TCI
Crystal violet
Catalog # TCIAC0428-100G
Supplier:  TCI
CAS Number:  548-62-9
Crystal violet
Crystal violet

100 g

CAS Number:  548-62-9
Price
476,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
  • CAS Number:548-62-9
  • Formula:C₂₅H₃₀ClN₃
  • MDL Number:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:14,04395
  • Melting Pt:211°C
  • EINECS:208-953-6
  • UN:3077
  • ADR:9,III
Specifications

Specification Test Results

  • Appearance:
    Green to Dark green powder to crystal
  • Absorbance:
    min. 0.300 (2mg/L, 588.0 to 592.0 nm) (calcd.on dried substance)
  • Ethanol insoluble matter:
    max. 20.0 %
  • Drying loss:
    max. 10.0 %
  • Sensitiveness:
    Turn Purple in Acetic Acid and turn Yellow green by adding Perchloric Acid

Specifications

  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.