TUBE AND IND CAPS 0.2ML PCR 8 STRIP ASS 1 * 120 items
Catalog # ALPCLW2510AS
Supplier: Alpha Laboratories
TUBE AND IND CAPS 0.2ML PCR 8 STRIP ASS 1 * 120 items
Catalog # ALPCLW2510AS
Supplier: Alpha Laboratories
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR®, Accessories for VWR® SuperClear Screw Cap Microcentrifuge Tubes
Supplier: VWR Collection
Screw cap microcentrifuge tubes, SuperClear®
Supplier: LABCON
Cryoboxes, 133×133 mm
Supplier: RATIOLAB
Cell Scraper, Lifter Blade, 39 cm 1 * 100 items
Supplier: STEMCELL Technologies
Nalgene® Square PETG Platinum Certified Clean Media Bottles
Supplier: Thermo Fisher Scientific
OmniTop Sample Tubes® Exact Volume Sampling System (EVSS)
Supplier: Avantor Fluid Handling
Collagen I-coated microplates, Corning® BioCoat®
Supplier: Corning
Coolers for Reagent Reservoirs
Supplier: DIVERSIFIED BIOTECH
Cryoboxes
Supplier: RATIOLAB
HUMAN IL-2 IS, RESEARCH GRADE 1 * 50 µG
Supplier: MILTENYI BIOTEC
PCR Thermo-Tube and cap strips, 0.2 ml, ABgene®
Supplier: Thermo Fisher Scientific
Microcentrifuge tubes, 1,5 ml, in resealable bags, SuperSpin®
Supplier: LABCON