Order Entry
Denmark
ContactUsLinkComponent
MICRO CYCLONIC SPRAY CHAMBERS PROVIDE THE FASTEST WASH-OUT AT LOW-FLOW RATES COMPARED TO LARGER VERSIONS. NARROW BAFFLE USED WITH OI LFAST AND ORGANIC SOLVENT APPLICATIONS. DO NOT USE FOR AQUEOUS APPLICATIONS. 1 * 1 items
Catalog # PERCN0777638
Supplier:  PerkinElmer
MICRO CYCLONIC SPRAY CHAMBERS PROVIDE THE FASTEST WASH-OUT AT LOW-FLOW RATES COMPARED TO LARGER VERSIONS. NARROW BAFFLE USED WITH OI LFAST AND ORGANIC SOLVENT APPLICATIONS. DO NOT USE FOR AQUEOUS APPLICATIONS. 1 * 1 items
Catalog # PERCN0777638
Supplier:  PerkinElmer
Frequently Bought Together