Order Entry
Denmark
ContactUsLinkComponent
F0 ENCAPSULATED MICRO TUBING FOR MICROFLOW RATES WITH THE LOWEST INTERNAL VOLUME OF ANY NEBULIZER. INCLUDES 0.25 MM I.D. (GREEN MARK ER) 70 CM LONG MICRO CAPILLARY. COMPATIBLE WITH ALL MEINHARD PLUS NEBULIZERS. 1 * 1 items
Catalog # PERCN8121038
Supplier:  PerkinElmer
F0 ENCAPSULATED MICRO TUBING FOR MICROFLOW RATES WITH THE LOWEST INTERNAL VOLUME OF ANY NEBULIZER. INCLUDES 0.25 MM I.D. (GREEN MARK ER) 70 CM LONG MICRO CAPILLARY. COMPATIBLE WITH ALL MEINHARD PLUS NEBULIZERS. 1 * 1 items
Catalog # PERCN8121038
Supplier:  PerkinElmer
Frequently Bought Together