Order Entry
Denmark
ContactUsLinkComponent
EACH COMPRESSOR COMES ASSEMBLED ON AN INTERNALLY AND EXTERNALLY COATED AIR TANK AND INCLUDES OPTIONAL SELF-PURGING REGENERATIVE AIR DRYER WITH AFTERCOOLER AND 5 MICRON PARTICLE FILTERS. ONE VERSION INCLUDES A SILENCING CABINET RESULTING IN A COMPRESS 1 * 1 items
Catalog # PERCN0777883
Supplier:  PerkinElmer
EACH COMPRESSOR COMES ASSEMBLED ON AN INTERNALLY AND EXTERNALLY COATED AIR TANK AND INCLUDES OPTIONAL SELF-PURGING REGENERATIVE AIR DRYER WITH AFTERCOOLER AND 5 MICRON PARTICLE FILTERS. ONE VERSION INCLUDES A SILENCING CABINET RESULTING IN A COMPRESS 1 * 1 items
Catalog # PERCN0777883
Supplier:  PerkinElmer
Frequently Bought Together