Pallets and sumps
Sump pallet, steel, type SW10, for 10×200 l drums, painted model, loading capacity 2800 kg, weight 175 kg, 3006×1205×266 mm
Catalog # CEMG10408
Supplier: CEMO
Jump to:
Product Details & Documents
The steel sumps provide safe storage for water-polluting as well as inflammable substances.
- All SW sump pallets are equipped with hot-dip galvanised, removable gratings
- A wide range of models ensures the correct solution for many application
- Sump pallets with 100 mm ground clearance permit driving under from all sides
Mobile steel sump pallets contains two steering and two fixed castors and a pushing bar for trouble-free steering.
Specifications
- Description:Sump pallet, steel, type SW10, for 10×200 l drums, painted model, loading capacity 2800 kg, weight 175 kg
- Seq:15
- W×D×H:3006×1205×266 mm
Accessories
Product Information
- Catalog #WeightW×D×HAvailabilityPrice
- Barrel support type FB3 for GRP and steel sump pallets, capacity: 3 x 60 lBarrel support type FB3 for GRP and steel sump pallets, capacity: 3 x 60 lCEMG783517 kg1200×600×400 mmLog in for availability1 * 1 Items - 1.630,00 kr
Product Information
- Catalog #WeightW×D×HAvailabilityPrice
- Barrel support stackable for GRP and steel sump pallets, capacity: 1 x 60 lBarrel support stackable for GRP and steel sump pallets, capacity: 1 x 60 lCEMG774110 kg630×450×530 mmLog in for availability1 * 1 Items - 1.940,00 kr
Product Information
- Catalog #WeightW×D×HAvailabilityPrice
- Barrel support type FB1 for GRP and steel sump pallets, capacity: 1 x 60 l or 1 x 200 lBarrel support type FB1 for GRP and steel sump pallets, capacity: 1 x 60 l or 1 x 200 lCEMG758615 kg600×600×350 mmLog in for availability1 * 1 Items - 1.160,00 kr
Product Information
- Catalog #WeightW×D×HAvailabilityPrice
- Barrel support type FB2 for GRP and steel sump pallets, capacity 2 x 60 l or 2 x 200 lBarrel support type FB2 for GRP and steel sump pallets, capacity 2 x 60 l or 2 x 200 lCEMG758721 kg1130×600×350 mmLog in for availability1 * 1 Items - 1.400,00 kr
Product Information
- Catalog #WeightW×D×HAvailabilityPrice
- Drum pallet, PE, for 2×200 l drumsDrum pallet, PE, for 2×200 l drumsCEMG867515 kg940×1380×390 mmLog in for availability1 * 1 Items - 2.100,00 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



