Hydrochloric acid fuming 37% for analysis EMPARTA® ACS 1 * 2,5 L
1.01834.2502
: Merck
Frequently Bought Together
Magnesium chloride, anhydrous ≥98%, high purity
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Qualitative Filter Papers
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Erlenmeyer flasks, narrow or wide neck, with beaded rim, DURAN®
: DWK Life Sciences
Ethylene glycol ≥98%, TECHNICAL
: VWR Chemicals
Disposable cuvettes
: Brand
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Glass microfibre filters without binder, grade GF/A, Whatman™
: Whatman products (Cytiva)
Titrants and reagents for volumetric Karl Fischer titrations, HYDRANAL™, Fluka™
: Honeywell Chemicals
Sodium sulphate, anhydrous ≥99.0%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
: Honeywell Chemicals