Order Entry
Denmark
Orders LinkContactUsLinkComponent
Microplate handling system, StakMax®
Microplate handling system, StakMax®
50-plate stacker magazines (pair)
Catalog # MLDV0200-6187
Supplier:   Molecular Devices
Microplate handling system, StakMax®
Catalog # MLDV0200-6187
Supplier:  Molecular Devices
Microplate handling system, StakMax®
Microplate handling system, StakMax®
50-plate stacker magazines (pair)
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
The StakMax® microplate handling system is a compact benchtop microplate stacker that works with microplate readers and microplate washers to give walk-away automation for up to 50 microplates.
  • Integrates with microplate readers and AquaMax® washers for improved process flow and higher throughput
  • Flexible operation using powerful SoftMax® Pro software
  • Barcode reader can be added to enable barcode identification of plates
  • Minimises plate movement time, while handling the microplates gently, eliminating liquid spillage
Specifications
  • Description:
    50-plate stacker magazines (pair)
  • Seq:
    07