Order Entry
Denmark
Orders LinkContactUsLinkComponent
Biological indicators, spore threads
Biological indicators, spore threads
Spore threads, individually packed in glassine envelopes
Catalog # NAMSTHS-06P
Supplier:   CROSSTEX
Biological indicators, spore threads
Catalog # NAMSTHS-06P
Supplier:  CROSSTEX
Biological indicators, spore threads
Spore threads, individually packed in glassine envelopes
Price
10.210,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
Spore threads are prepared from Geobacillus stearothermophilus. These are designed to be placed directly into a device and utilised to monitor steam sterilisation process efficacy.
Ideal for use in tubing, lumens and other devices/instruments where larger BIs and carriers cannot be utilised.
Specifications
  • Description:
    Spore threads, individually packed in glassine envelopes