Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Rotary evaporating flasks

1000 ml, 24/29 NS, 204 mm
  BIBBFD1L/3
 :   SciLabware
Rotary evaporating flasks
  BIBBFD1L/3
Rotary evaporating flasks
Rotary evaporating flasks

1000 ml, 24/29 NS, 204 mm

 
3.280,00 kr
   
1 * 5 Items
 

PYREX®, borosilicate glass.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Ideal for use with rotary evaporators. Large surface area for rapid evaporation. Robust heavy wall construction for added strength. Florentine shape allows easy removal of solid residues.

 
  • Socket size:
    24/29 NS
  • Capacity:
    1000 ml
  • Seq:
    10
  • Total height:
    204 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.