Rotary evaporating flasks
500 ml, 24/29 NS, 165 mm
Catalog # BIBBFD500/3
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for use with rotary evaporators. Large surface area for rapid evaporation. Robust heavy wall construction for added strength. Florentine shape allows easy removal of solid residues.
Specifications
- Socket size:24/29 NS
- Capacity:500 ml
- Seq:08
- Total height:165 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



