Order Entry
Denmark
ContactUsLinkComponent
made of stainless steel Ideal for VisiCon safety containers. Opens by pushing or pulling. Self-closing prevents uncontrolled leakage of liquids. Setting ring for adjustment, after installation vertically adjustable. 1 * 1 items
Catalog # DUEH5.88.100
Supplier:  DUPERTHAL
made of stainless steel Ideal for VisiCon safety containers. Opens by pushing or pulling. Self-closing prevents uncontrolled leakage of liquids. Setting ring for adjustment, after installation vertically adjustable. 1 * 1 items
Catalog # DUEH5.88.100
Supplier:  DUPERTHAL
Frequently Bought Together