Order Entry
Denmark
Orders LinkContactUsLinkComponent
Apigenin ≥98% Anticancer agent

Apigenin ≥98% Anticancer agent

Catalog # ABCAAB120141-10
Marketsource
Supplier:   Abcam
Apigenin ≥98% Anticancer agent
Catalog # ABCAAB120141-10
Supplier:  Abcam
Marketsource
CAS Number:  520-36-5
Apigenin ≥98% Anticancer agent
CAS Number:  520-36-5
Price
510,00 kr
Log in for availability
1 * 10 mg
Product Details & Documents

Plant flavone. Antioxidant, anti-infla mMatory and anticancer properties. Effects a wide range of molecular targets including caspases, fatty acid synthase, topoisomerase and nuclear factor-κB.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  4',5,7-Trihydroxyflavone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4',6'-Dihydroxy-2-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
  • CAS Number:520-36-5
  • Molecular Weight:270,24 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% 5,7-dihydroxy-2-(4-hydroxyphenyl)-4-benzopyrone
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.