Order Entry
Denmark
ContactUsLinkComponent
LDPE bags, clear, compatible for use in cleanroom. Thickness 2mil (50µm), Width 10 (254mm), Length 40 (1016mm). Cleanliness level 100 as per NASA JPAG 5322.1 100 pcs per double-pack 1 * 100 items
Catalog # KNFCLB1021040
Supplier:  KNF CLEANROOM
LDPE bags, clear, compatible for use in cleanroom. Thickness 2mil (50µm), Width 10 (254mm), Length 40 (1016mm). Cleanliness level 100 as per NASA JPAG 5322.1 100 pcs per double-pack 1 * 100 items
Catalog # KNFCLB1021040
Supplier:  KNF CLEANROOM