Worktables
Stainless steel, 1800×700×900 mm
Catalog # HUPF75R60302-1800
Supplier: HUPFER METALLWERKE
Jump to:
Product Details & Documents
This open version work table features closed table top and bottom side covered with 0,8 mm sheet steel.
- Particle board stuffing
- Sandwich panelled table top
Working table made of stainless steel with grain size 400 ground and material thickness of 1 mm.
Documents
HUPF75R60302-1800_TDS_EN
Datasheet
Specifications
- Material:Stainless steel
- W×D×H:1800×700×900 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



