Order Entry
Denmark
Orders LinkContactUsLinkComponent
EDTA solutions

EDTA solutions

EDTA standard solution, 0,0075N, 100 ml marked dropping bottle, 0 - 360 ka/ha, Gypsum requirement drop count titration, 100 mL
Catalog # HACH2498132
Supplier:   Hach
EDTA solutions
Catalog # HACH2498132
Supplier:  Hach
CAS Number:  
EDTA solutions
EDTA solutions

EDTA standard solution, 0,0075N, 100 ml marked dropping bottle, 0 - 360 ka/ha, Gypsum requirement drop count titration, 100 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Chemical method:
    Gypsum requirement drop count titration
  • Pk:
    100 mL
  • Description:
    EDTA standard solution, 0,0075N, 100 ml marked dropping bottle
  • Range:
    0 - 360 ka/ha

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.