Order Entry
Denmark
ContactUsLinkComponent
2,2-AZINO-bis-(3-ETHYLBENZTHIAZOLINE-6-SULFONIC ACID) SOLUTION. Stabilized Liquid SubstrateA safe, convenient and good overall performing substrate for ELISA. It may be used for both kinetic and endpoint reactions. Supplied ready-to-use at 1.46 mmol 1 * 100 mL
Catalog # ICNA04821809
Supplier:  MP Biomedicals
2,2-AZINO-bis-(3-ETHYLBENZTHIAZOLINE-6-SULFONIC ACID) SOLUTION. Stabilized Liquid SubstrateA safe, convenient and good overall performing substrate for ELISA. It may be used for both kinetic and endpoint reactions. Supplied ready-to-use at 1.46 mmol 1 * 100 mL
Catalog # ICNA04821809
Supplier:  MP Biomedicals
Frequently Bought Together