Order Entry
Denmark
ContactUsLinkComponent
CALYCULIN A. Purity: >95%Phosphatase inhibitor with very potent inhibitory activity against both PP1 and PP2A phosphatases.IC50 valu es of approximately 1 nm for both PP1 and PP2A. 1 * 10 µG
Catalog # ICNA0215837810
Supplier:  MP Biomedicals
CALYCULIN A. Purity: >95%Phosphatase inhibitor with very potent inhibitory activity against both PP1 and PP2A phosphatases.IC50 valu es of approximately 1 nm for both PP1 and PP2A. 1 * 10 µG
Catalog # ICNA0215837810
Supplier:  MP Biomedicals
Frequently Bought Together