Order Entry
Denmark
Orders LinkContactUsLinkComponent
Paraformaldehyde, powder

Paraformaldehyde, powder

Glass bottle for solids, 1 kg
Catalog # ICNA0215014601
Supplier:   MP Biomedicals
Paraformaldehyde, powder
Catalog # ICNA0215014601
Supplier:  MP Biomedicals
CAS Number:  30525-89-4
Paraformaldehyde, powder

Glass bottle for solids, 1 kg

CAS Number:  30525-89-4
Price
893,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents

Soluble in water (hot water - ~37% w/v; slowly soluble in cold water). Aqueous solutions will develop formaldehyde over time. Insoluble in alcohol or ether.

Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • UN:2213
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.