3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate
100 mL
Catalog # ICNA0215234680
Supplier: MP Biomedicals
3,3',5,5'-Tetramethylbenzidine (TMB), liquid substrate
Catalog # ICNA0215234680
Supplier: MP Biomedicals
CAS Number: 54827-17-7
Jump to:
Product Details & Documents
TMB is a chromogenic substrate for Horse Radish Peroxidase (HRP). MP Biolomedicals TMB Solution is a convenient, non toxic and ready to use stable chromogen solution suitable for peroxidase and pseudo-peroxidase reactions in ELISA, it provides consistent performance and sensitivity to deliver reproducible results
- Ready-to-Use, convenient
- Reproducible Results
- Consistent Performance and Sensitivity
- Stabilized chromogen solution
- For ELISA procedures and peroxidase and pseudo-peroxidase reactions
- Stable up to 24 months
- Non toxic.
Metabolism of the TMB substrate by HRP creates a soluble bluish reaction product that may be quantified by measuring absorbance at 370 or 665 nm. Reaction can be stopped by addition of 1 volume of 1N hydrocloric or sulfurc acid to yield a yellow reaction product that can be measured by absorbance at 450 nm (within one hour).
Caution: Research Use Only (RUO).
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
- CAS Number:54827-17-7
- Formula:C₁₆H₂₀N₂
- MDL Number:MFCD00007748
- Molecular Weight:240.35 g/mol
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Identity Test:Passes
- Functionality Test :Passes
- pH:3.1 ± 0.5
- Expiration Date:Month/Day/Year
Specifications
- Pk:100 mL



