Order Entry
Denmark
Orders LinkContactUsLinkComponent
Lecithin

Lecithin

2,3 kg
Katalog # ICNA0210214805
Leverandør:   MP Biomedicals
Lecithin
Katalog # ICNA0210214805
Leverandør:  MP Biomedicals
CAS nummer:  8002-43-5
Lecithin

2,3 kg

CAS nummer:  8002-43-5
Price
1.280,00 kr
Log in for availability
1 * 2,3 kg
Product Details & Documents

Lecithin is a good source of choline, which may help with brain function and growth. Used as an emulsifier and solubilizer for drugs in an aqueous solution, also as a vehicle for liposomes. Lecithin is a phosphatidylcholine of soybean origin.

Storage temperature: -20 °C

  • CAS Number:8002-43-5
  • MDL Number:MFCD00147406
Specifications

Specifications

  • Pk:
    2,3 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.