Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Mannitol ≥96%, powder
500 g
  ICNA02102248.5
D-(-)-Mannitol ≥96%, powder
  ICNA02102248.5
 :  69-65-8
D-(-)-Mannitol ≥96%, powder
D-(-)-Mannitol ≥96%, powder
500 g
 :  69-65-8
536,00 kr
1 * 500 g
Mannitol is carbohydrate found in plants and plant exudates.
A sugar alcohol sweet tastant. Used in sweetness inhibition studies.
A sugar alcohol sweet tastant. Used in sweetness inhibition studies.
Soluble in water (1 g/5,5 ml), alcohol (1 g/83 ml), glycerol (1 g/18 ml); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.
 :  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  •  :69-65-8
  •  :HOH₂C(CH(OH))₄CH₂OH
  •  :MFCD00064287
  •  :182.17 g/mol
  •  :13,05769
  •  :290…295 °C (4 hPa)
  •  :163…172 °C
  •  :1.49 g/cm³ (20 °C)
  •  :Ambient
  •  :200-711-8
  • Pk:
    500 g