Order Entry
Denmark
ContactUsLinkComponent
CHYMOTRYPSIN. From Human Pancreas E.C.3.4.21.1.Activity: 75-95% active sites by burst titrant analysis.Purity: >99% by SDS-PAGE. Supplied as a lyophilized powder from 1 mM hydrochloric acid. 1 * 100 µG
Catalog # ICNA0219133880
Supplier:  MP Biomedicals
CHYMOTRYPSIN. From Human Pancreas E.C.3.4.21.1.Activity: 75-95% active sites by burst titrant analysis.Purity: >99% by SDS-PAGE. Supplied as a lyophilized powder from 1 mM hydrochloric acid. 1 * 100 µG
Catalog # ICNA0219133880
Supplier:  MP Biomedicals
Frequently Bought Together